Assay Details
| Gene Symbol: Ccl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019577.4 | NP_062523.1 | 2 - 3 | 264 | 95 | |
| GenBank mRNA | AF244367.1 | - | 2 - 3 | 193 | 95 | |
| AF281075.1 | - | 2 - 3 | 246 | 95 | ||
| AK009718.1 | - | 2 - 3 | 236 | 95 | ||
| AK157068.1 | - | 2 - 3 | 265 | 95 | ||
| BC065389.1 | - | 2 - 3 | 270 | 3472002 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of endothelial cell proliferation
chemotaxis
inflammatory response
immune response
cytoskeleton organization
regulation of cell shape
positive regulation of cell migration
positive regulation of actin filament polymerization
positive regulation of GTPase activity
positive regulation of angiogenesis
eosinophil chemotaxis
positive regulation of inflammatory response
positive regulation of eosinophil migration
cytokine activity
chemokine activity
positive regulation of endothelial cell proliferation
chemotaxis
inflammatory response
immune response
cytoskeleton organization
regulation of cell shape
positive regulation of cell migration
positive regulation of actin filament polymerization
positive regulation of GTPase activity
positive regulation of angiogenesis
eosinophil chemotaxis
positive regulation of inflammatory response
positive regulation of eosinophil migration
cytokine activity
chemokine activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322640 | NM_019577.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322640 | AF244367.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322640 | AF281075.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322640 | AK009718.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322640 | AK157068.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322640 | NM_019577.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322640 | AF244367.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322640 | AF281075.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322640 | AK009718.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P322640 | AK157068.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 383485 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 761301 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 77505 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147150 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P322640 | NM_019577.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P322640 | AF244367.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P322640 | AF281075.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P322640 | AK009718.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P322640 | AK157068.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P322640 | NM_019577.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P322640 | AF244367.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P322640 | AF281075.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P322640 | AK009718.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P322640 | AK157068.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P322640 | NM_019577.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P322640 | AF244367.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P322640 | AF281075.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P322640 | AK009718.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P322640 | AK157068.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P322640 | NM_019577.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P322640 | AF244367.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P322640 | AF281075.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P322640 | AK009718.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P322640 | AK157068.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P18116 | NM_019577.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P18116 | AF281075.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P18116 | AK009718.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P18116 | AK157068.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116915 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116911 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116902 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116922 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116913 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116905 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116918 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116935 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116920 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24116934 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147150 | - | - |
| Whole Mouse Genome G4122A | A_51_P322640 | NM_019577.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P322640 | AF244367.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P322640 | AF281075.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P322640 | AK009718.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P322640 | AK157068.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P322640 | NM_019577.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P322640 | AF244367.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P322640 | AF281075.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P322640 | AK009718.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P322640 | AK157068.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P18116 | NM_019577.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P18116 | AF281075.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P18116 | AK009718.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P18116 | AK157068.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137664 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139777 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147150 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04145230 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04214747 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04214752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24116884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116935 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04866458 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04775337 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116941 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04437646 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04859649 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116911 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04204339 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04126180 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04153455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116938 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04975550 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116909 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04214752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116934 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116943 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04204333 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24116950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04170180 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04548081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04689289 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10534499 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534496 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534500 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534497 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534495 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534494 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534498 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450488_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10534493 | - | - |
| Mouse Genome 430 2.0 Array | 1450488_at | - | - |
| Mouse Genome 430 2.0 Array | 1450488_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56221_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56221_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500030726.mm.1 | TC0500003399.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500030726.mm.1 | NM_019577.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500030726.mm.1 | NM_019577.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500030729.mm.1 | TC0500003399.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500030729.mm.1 | NM_019577.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500030729.mm.1 | NM_019577.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500030728.mm.1 | TC0500003399.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500030728.mm.1 | NM_019577.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500030728.mm.1 | NM_019577.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500030727.mm.1 | TC0500003399.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500030727.mm.1 | NM_019577.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500030727.mm.1 | NM_019577.4 | Refseq |
| Mouse Expression Set 430 | 1450488_PM_at | - | - |
| Mouse Expression Set 430 | 1450488_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450488_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450488_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450488_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450488_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 349163 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 670129 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000670129 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225406 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225406 | - | - |
| mouse-6 v1.1 (Illumina) | scl25923.3.1_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 670129 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl25923.3.1_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 670129 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1225406 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1225406 | - | - |
Citations (2)
Foxa2 programs Th2 cell-mediated innate immunity in the developing lung.
- Authors
- Chen G, Wan H, Luo F, Zhang L, Xu Y, Lewkowich I, Wills-Karp M, Whitsett JA
- Journal
- J Immunol - Page No 6133 (Tue Jun 01 00:00:00 EDT 2010)
- Volume (Year)
- 184 (2010)
- Product Usage
- -
IL-21 administration into the nostril alleviates murine allergic rhinitis.
- Authors
- Hiromura Y, Kishida T, Nakano H, Hama T, Imanishi J, Hisa Y, Mazda O
- Journal
- J Immunol - Page No 7157 (Thu Nov 15 00:00:00 EST 2007)
- Volume (Year)
- 179 (2007)
- Product Usage
- -

Ccl24
Genomic Map