Assay Details
| Gene Symbol: Srpk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019684.1 | NP_062658.1 | 5 - 6 | 504 | 65 | |
| GenBank mRNA | AF043288.1 | - | 5 - 6 | 504 | 65 | |
| AJ748651.1 | - | 4 - 5 | 328 | 65 | ||
| AK004696.2 | - | 5 - 6 | 522 | 65 | ||
| BC120883.1 | - | 5 - 6 | 499 | 40057934 | 65 | |
| BC120884.1 | - | 5 - 6 | 499 | 40057937 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Srpk3
Genomic Map