Assay Details
| Gene Symbol: Srpk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019684.1 | NP_062658.1 | 6 - 7 | 611 | 81 | |
| GenBank mRNA | AF043288.1 | - | 6 - 7 | 611 | 81 | |
| AJ748651.1 | - | 5 - 6 | 435 | 81 | ||
| BC120883.1 | - | 6 - 7 | 606 | 40057934 | 81 | |
| BC120884.1 | - | 6 - 7 | 606 | 40057937 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Srpk3
Genomic Map