Assay Details
| Gene Symbol: Srpk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019684.1 | NP_062658.1 | 8 - 9 | 798 | 83 | |
| GenBank mRNA | AF043288.1 | - | 8 - 9 | 798 | 83 | |
| AJ748651.1 | - | 7 - 8 | 622 | 83 | ||
| AK003681.1 | - | 2 - 3 | 160 | 83 | ||
| BC120883.1 | - | 8 - 9 | 793 | 40057934 | 83 | |
| BC120884.1 | - | 8 - 9 | 793 | 40057937 | 83 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
multicellular organism development
muscle organ development
skeletal muscle tissue development
phosphorylation
cell differentiation
muscle tissue development
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein phosphorylation
multicellular organism development
muscle organ development
skeletal muscle tissue development
phosphorylation
cell differentiation
muscle tissue development
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P375862 | AK004696.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P375862 | BC120883.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P375862 | BC120884.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P375862 | NM_019684.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P375862 | AF043288.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P375862 | AK004696.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P375862 | BC120883.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P375862 | BC120884.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P375862 | NM_019684.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P375862 | AF043288.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 274223 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 90665 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 726017 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 573888 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P176771 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P375862 | AK004696.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P375862 | BC120883.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P375862 | BC120884.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P375862 | NM_019684.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P375862 | AF043288.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P375862 | AK004696.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P375862 | BC120883.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P375862 | BC120884.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P375862 | NM_019684.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P375862 | AF043288.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464649 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464682 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464640 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464667 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464653 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464665 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464645 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464664 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464677 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464654 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464632 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464651 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464638 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464631 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464680 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464636 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464646 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464635 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464683 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32464660 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P176771 | - | - |
| Whole Mouse Genome G4122A | A_51_P375862 | AK004696.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P375862 | BC120883.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P375862 | BC120884.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P375862 | NM_019684.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P375862 | AF043288.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P375862 | AK004696.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P375862 | BC120883.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P375862 | BC120884.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P375862 | NM_019684.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P375862 | AF043288.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118785 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139449 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P176771 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15192063 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15192026 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15192033 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15192000 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15192075 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15192039 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15192067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32464689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464679 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464660 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464693 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15192005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15192039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464663 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464673 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15192075 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15192074 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464696 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464665 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464685 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464697 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464662 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15192001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464659 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464677 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15192099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464703 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464674 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464664 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464694 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464698 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32464670 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10600292 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600286 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600296 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600299 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600290 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600288 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600298 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600297 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600300 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600291 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600295 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600293 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600289 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600285 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600294 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600287 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1432139_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418798_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1447806_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431825_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10600284 | - | - |
| Mouse Genome 430 2.0 Array | 1418798_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1447806_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1447806_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1431825_at | - | - |
| Mouse Genome 430 2.0 Array | 1431825_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1432139_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418798_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1432139_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56504_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56504_at | - | - |
| Mouse Expression Set 430 | 1432139_PM_at | - | - |
| Mouse Expression Set 430 | 1432139_at | - | - |
| Mouse Expression Set 430 | 1418798_s_at | - | - |
| Mouse Expression Set 430 | 1418798_PM_s_at | - | - |
| Mouse Expression Set 430 | 1431825_at | - | - |
| Mouse Expression Set 430 | 1447806_PM_s_at | - | - |
| Mouse Expression Set 430 | 1447806_s_at | - | - |
| Mouse Expression Set 430 | 1431825_PM_at | - | - |
| Murine Genome U74v2 Set | 92436_PM_at | - | - |
| Murine Genome U74v2 Set | 92436_at | - | - |
| Mouse Genome 430A 2.0 Array | 1432139_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418798_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418798_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1432139_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418798_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418798_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1432139_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1432139_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447806_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431825_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447806_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431825_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 767779 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2480278 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002480278 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2684515 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2684515 | - | - |
| mouse-6 v1.1 (Illumina) | scl056504.15_296-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2480278 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl056504.15_296-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2480278 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2684515 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2684515 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl056504.15_296-S | - | - |

Srpk3
Genomic Map