Assay Details
| Gene Symbol: Srpk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019684.1 | NP_062658.1 | 9 - 10 | 931 | 69 | |
| GenBank mRNA | AF043288.1 | - | 9 - 10 | 931 | 69 | |
| AJ748651.1 | - | 8 - 9 | 755 | 69 | ||
| AK003681.1 | - | 3 - 4 | 293 | 69 | ||
| AK004696.2 | - | 6 - 7 | 1600 | 69 | ||
| BC120883.1 | - | 9 - 10 | 926 | 40057934 | 69 | |
| BC120884.1 | - | 9 - 10 | 926 | 40057937 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Srpk3
Genomic Map