Assay Details
| Gene Symbol: Aldh18a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019698.2 | NP_062672.2 | 10 - 11 | 1323 | 96 | |
| NM_153554.2 | NP_705782.2 | 10 - 11 | 1317 | 96 | ||
| GenBank mRNA | AF056573.1 | - | 10 - 11 | 1228 | 96 | |
| AF056574.1 | - | 10 - 11 | 1222 | 96 | ||
| AK134148.1 | - | 10 - 11 | 1283 | 96 | ||
| AK147711.1 | - | 10 - 11 | 1243 | 96 | ||
| AK151072.1 | - | 10 - 11 | 1245 | 96 | ||
| AK153946.1 | - | 10 - 11 | 1244 | 96 | ||
| AK159610.1 | - | 10 - 11 | 1245 | 96 | ||
| AK166459.1 | - | 10 - 11 | 1209 | 96 | ||
| AK166494.1 | - | 10 - 11 | 1210 | 96 | ||
| AK168905.1 | - | 10 - 11 | 1243 | 96 | ||
| AK200973.1 | - | 1 - 2 | 60 | 96 | ||
| AK202353.1 | - | 62 | 97 | |||
| BC033427.1 | - | 10 - 11 | 1254 | 5009115 | 96 | |
| BC037699.1 | - | 10 - 11 | 1202 | 4188900 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldh18a1
Genomic Map