Assay Details
| Gene Symbol: Aldh18a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019698.2 | NP_062672.2 | 15 - 16 | 2049 | 59 | |
| NM_153554.2 | NP_705782.2 | 15 - 16 | 2043 | 59 | ||
| GenBank mRNA | AF056573.1 | - | 15 - 16 | 1954 | 59 | |
| AF056574.1 | - | 15 - 16 | 1948 | 59 | ||
| AK134148.1 | - | 15 - 16 | 2009 | 59 | ||
| AK147711.1 | - | 15 - 16 | 1969 | 59 | ||
| AK151072.1 | - | 15 - 16 | 1971 | 59 | ||
| AK153946.1 | - | 15 - 16 | 1970 | 59 | ||
| AK159610.1 | - | 15 - 16 | 1971 | 59 | ||
| AK168905.1 | - | 15 - 16 | 1969 | 59 | ||
| BC033427.1 | - | 15 - 16 | 1980 | 5009115 | 59 | |
| BC037699.1 | - | 15 - 16 | 1928 | 4188900 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldh18a1
Genomic Map