Assay Details
| Gene Symbol: Arl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019718.2 | NP_062692.1 | 2 - 3 | 187 | 106 | |
| GenBank mRNA | AF143241.1 | - | 1 - 2 | 144 | 106 | |
| AK048644.1 | - | 2 - 3 | 195 | 106 | ||
| BC042941.1 | - | 2 - 3 | 187 | 6493571 | 106 | |
More Information
Set Membership: |
|

Arl3
Genomic Map