Assay Details
| Gene Symbol: Arl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019718.2 | NP_062692.1 | 4 - 5 | 359 | 72 | |
| GenBank mRNA | AF143241.1 | - | 3 - 4 | 316 | 72 | |
| AK048644.1 | - | 4 - 5 | 367 | 72 | ||
| BC042941.1 | - | 4 - 5 | 359 | 6493571 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arl3
Genomic Map