Assay Details
| Gene Symbol: Nme4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019731.1 | NP_062705.1 | 3 - 4 | 342 | 93 | |
| GenBank mRNA | AF153451.1 | - | 3 - 4 | 342 | 93 | |
| AF288690.1 | - | 3 - 4 | 355 | 93 | ||
| AK012805.1 | - | 3 - 4 | 378 | 93 | ||
| AK017724.1 | - | 3 - 4 | 358 | 93 | ||
More Information
Set Membership: |
|

Nme4
Genomic Map