Assay Details
| Gene Symbol: Dnaja2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019794.4 | NP_062768.1 | 1 - 2 | 104 | 77 | |
| GenBank mRNA | AB028853.1 | - | 1 - 2 | 105 | 77 | |
| AK077672.1 | - | 1 - 2 | 175 | 77 | ||
| AK083208.1 | - | 1 - 2 | 172 | 77 | ||
| AK163255.1 | - | 1 - 2 | 103 | 77 | ||
| AK169169.1 | - | 1 - 2 | 164 | 77 | ||
| BC003420.1 | - | 1 - 2 | 177 | 2582183 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnaja2
Genomic Map