Assay Details
| Gene Symbol: Dnaja2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019794.4 | NP_062768.1 | 4 - 5 | 464 | 97 | |
| GenBank mRNA | AB028853.1 | - | 4 - 5 | 465 | 97 | |
| AK031858.1 | - | 4 - 5 | 531 | 97 | ||
| AK077672.1 | - | 4 - 5 | 535 | 97 | ||
| AK083208.1 | - | 4 - 5 | 532 | 97 | ||
| AK163255.1 | - | 4 - 5 | 463 | 97 | ||
| AK169169.1 | - | 4 - 5 | 524 | 97 | ||
| AK212721.1 | - | 2 - 3 | 175 | 97 | ||
| BC003420.1 | - | 4 - 5 | 537 | 2582183 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnaja2
Genomic Map