Assay Details
| Gene Symbol: Dnaja2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019794.4 | NP_062768.1 | 7 - 8 | 941 | 77 | |
| GenBank mRNA | AB028853.1 | - | 7 - 8 | 942 | 77 | |
| AK031858.1 | - | 7 - 8 | 1008 | 77 | ||
| AK077672.1 | - | 7 - 8 | 1012 | 77 | ||
| AK083208.1 | - | 7 - 8 | 1009 | 77 | ||
| AK163255.1 | - | 7 - 8 | 940 | 77 | ||
| AK169169.1 | - | 7 - 8 | 1001 | 77 | ||
| AK211376.1 | - | 1 - 2 | 163 | 77 | ||
| BC003420.1 | - | 7 - 8 | 1014 | 2582183 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnaja2
Genomic Map