Assay Details
| Gene Symbol: Dclk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111051.1 | NP_001104521.1 | 9 - 10 | 1153 | 92 | |
| NM_001111052.1 | NP_001104522.1 | 9 - 10 | 1153 | 92 | ||
| NM_001195538.1 | NP_001182467.1 | 13 - 14 | 2014 | 92 | ||
| NM_001195539.1 | NP_001182468.1 | 9 - 10 | 1201 | 92 | ||
| NM_019978.3 | NP_064362.1 | 13 - 14 | 2062 | 92 | ||
| GenBank mRNA | AF155819.1 | - | 13 - 14 | 1960 | 92 | |
| AF155821.1 | - | 9 - 10 | 1012 | 92 | ||
| AK032424.1 | - | 9 - 10 | 1043 | 92 | ||
| AK047716.1 | - | 9 - 10 | 940 | 92 | ||
| AK081200.1 | - | 9 - 10 | 996 | 92 | ||
| AY968047.1 | - | 9 - 10 | 901 | 92 | ||
| AY968048.1 | - | 9 - 10 | 904 | 92 | ||
| BC133685.1 | - | 13 - 14 | 1816 | 40110683 | 92 | |
| BQ930085.1 | - | 1 - 2 | 89 | 92 | ||
More Information
Set Membership: |
|

Dclk1
Genomic Map