Assay Details
| Gene Symbol: Dkk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020265.4 | NP_064661.2 | 1 - 2 | 968 | 78 | |
| GenBank mRNA | AJ243963.2 | - | 1 - 2 | 368 | 78 | |
| AK028225.1 | - | 1 - 2 | 253 | 78 | ||
| AK031749.1 | - | 1 - 2 | 969 | 78 | ||
| BC096448.1 | - | 1 - 2 | 959 | 6389874 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dkk2
Genomic Map