Assay Details
| Gene Symbol: Ank | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020332.4 | NP_065065.3 | 3 - 4 | 758 | 64 | |
| GenBank mRNA | AF001533.2 | - | 1 - 2 | 104 | 64 | |
| AF274752.1 | - | 3 - 4 | 461 | 64 | ||
| AK083135.1 | - | 3 - 4 | 759 | 64 | ||
| AK148070.1 | - | 3 - 4 | 531 | 64 | ||
| BC042644.1 | - | 3 - 4 | 683 | 64 | ||
| BC054379.1 | - | 3 - 4 | 665 | 6390764 | 64 | |
| DQ832285.1 | - | 3 - 4 | 758 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ank
Genomic Map