Assay Details
| Gene Symbol: Ank | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020332.4 | NP_065065.3 | 9 - 10 | 1467 | 76 | |
| GenBank mRNA | AF001533.2 | - | 7 - 8 | 813 | 76 | |
| AF274752.1 | - | 9 - 10 | 1170 | 76 | ||
| AK083135.1 | - | 9 - 10 | 1468 | 76 | ||
| AK086014.1 | - | 2 - 3 | 338 | 76 | ||
| BC042644.1 | - | 9 - 10 | 1390 | 76 | ||
| BC054379.1 | - | 9 - 10 | 1374 | 6390764 | 76 | |
| DQ832285.1 | - | 9 - 10 | 1467 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ank
Genomic Map