Assay Details
| Gene Symbol: Slc16a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020516.2 | NP_065262.1 | 4 - 5 | 555 | 69 | |
| GenBank mRNA | AF019111.2 | - | 4 - 5 | 564 | 69 | |
| BC018216.1 | - | 4 - 5 | 555 | 4481698 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc16a8
Genomic Map