Assay Details
| Gene Symbol: Tk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021028.3 | NP_066356.3 | 4 - 5 | 325 | 71 | |
| NR_045642.1 | 4 - 5 | 325 | 71 | |||
| GenBank mRNA | AF105217.1 | - | 4 - 5 | 300 | 71 | |
| AJ249341.2 | - | 4 - 5 | 303 | 71 | ||
| AK041651.1 | - | 4 - 5 | 317 | 71 | ||
| AK051286.1 | - | 4 - 5 | 319 | 71 | ||
| AK087557.1 | - | 4 - 5 | 326 | 71 | ||
| AK154160.1 | - | 4 - 5 | 319 | 71 | ||
| BC019982.1 | - | 4 - 5 | 317 | 4159742 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tk2
Genomic Map