Assay Details
| Gene Symbol: Cftr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021050.2 | NP_066388.1 | 5 - 6 | 715 | 131 | |
| GenBank mRNA | AK033621.1 | - | 5 - 6 | 633 | 131 | |
| M60493.1 | - | 5 - 6 | 716 | 131 | ||
| M69298.1 | - | 5 - 6 | 714 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cftr
Genomic Map