Assay Details
| Gene Symbol: Cftr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021050.2 | NP_066388.1 | 21 - 22 | 3596 | 138 | |
| GenBank mRNA | M60493.1 | - | 21 - 22 | 3597 | 138 | |
| M69298.1 | - | 21 - 22 | 3595 | 138 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cftr
Genomic Map