Assay Details
| Gene Symbol: Il4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021283.2 | NP_067258.1 | 1 - 2 | 189 | 63 | |
| GenBank mRNA | AB174765.1 | - | 1 - 2 | 168 | 63 | |
| AF352783.1 | - | 1 - 2 | 63 | |||
| BC027514.1 | - | 1 - 2 | 168 | 3376587 | 63 | |
| M13238.1 | - | 1 - 2 | 185 | 63 | ||
| M25892.1 | - | 1 - 2 | 217 | 63 | ||
| X03532.1 | - | 1 - 2 | 191 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il4
Genomic Map