Assay Details
| Gene Symbol: Slc7a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199015.1 | NP_001185944.1 | 4 - 5 | 447 | 54 | |
| NM_001199016.1 | NP_001185945.1 | 3 - 4 | 335 | 54 | ||
| NM_021291.3 | NP_067266.1 | 3 - 4 | 562 | 54 | ||
| GenBank mRNA | AA104782.1 | - | 3 - 4 | 396 | 54 | |
| AF192310.1 | - | 4 - 5 | 447 | 54 | ||
| AJ249198.1 | - | 3 - 4 | 562 | 54 | ||
| AK142541.1 | - | 3 - 4 | 336 | 54 | ||
| AK164714.1 | - | 3 - 4 | 334 | 54 | ||
| BC010746.1 | - | 4 - 5 | 425 | 4236578 | 54 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc7a9
Genomic Map