Assay Details
| Gene Symbol: Sec61a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021305.3 | NP_067280.1 | 6 - 7 | 550 | 68 | |
| GenBank mRNA | AF145254.1 | - | 6 - 7 | 486 | 68 | |
| AF222748.1 | - | 6 - 7 | 487 | 68 | ||
| AK017615.1 | - | 6 - 7 | 532 | 68 | ||
| AK077701.1 | - | 6 - 7 | 550 | 68 | ||
More Information
Set Membership: |
|

Sec61a2
Genomic Map