Assay Details
| Gene Symbol: Glp1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021332.2 | NP_067307.2 | 8 - 9 | 895 | 92 | |
| GenBank mRNA | AJ001692.1 | - | 8 - 9 | 885 | 92 | |
| BC103624.1 | - | 8 - 9 | 884 | 92 | ||
| BC105649.1 | - | 8 - 9 | 884 | 92 | ||
| BC105650.1 | - | 8 - 9 | 884 | 92 | ||
| BC139464.1 | - | 8 - 9 | 922 | 8862486 | 92 | |
| BC139466.1 | - | 8 - 9 | 922 | 8862488 | 92 | |
| BC145626.1 | - | 8 - 9 | 922 | 9054194 | 92 | |
| DQ093397.1 | - | 8 - 9 | 895 | 92 | ||
| DQ093398.1 | - | 8 - 9 | 895 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Glp1r
Genomic Map