Assay Details
| Gene Symbol: Slc26a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021353.3 | NP_067328.1 | 10 - 11 | 1428 | 85 | |
| GenBank mRNA | AF136751.1 | - | 10 - 11 | 1346 | 85 | |
| BC139271.1 | - | 9 - 10 | 1263 | 8862293 | 85 | |
| BC139273.1 | - | 9 - 10 | 1263 | 8862295 | 85 | |
More Information
Set Membership: |
|

Slc26a3
Genomic Map