Assay Details
| Gene Symbol: Itgb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159564.1 | NP_001153036.1 | 9 - 10 | 1255 | 99 | |
| NM_021359.3 | NP_067334.1 | 6 - 7 | 1149 | 99 | ||
| GenBank mRNA | AF115376.1 | - | 9 - 10 | 1101 | 99 | |
| AK008867.1 | - | 6 - 7 | 1148 | 99 | ||
| AK036439.1 | - | 9 - 10 | 1256 | 99 | ||
| AK085900.1 | - | 9 - 10 | 1296 | 99 | ||
| BC049185.1 | - | 6 - 7 | 1118 | 6475423 | 99 | |
More Information
Set Membership: |
|

Itgb6
Genomic Map