Assay Details
| Gene Symbol: Dnaja4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021422.4 | NP_067397.1 | 4 - 5 | 819 | 92 | |
| GenBank mRNA | AB032401.1 | - | 5 - 6 | 686 | 92 | |
| AK016666.1 | - | 3 - 4 | 416 | 92 | ||
| AK046476.1 | - | 4 - 5 | 820 | 92 | ||
| AK076175.1 | - | 4 - 5 | 820 | 92 | ||
| AK141909.1 | - | 4 - 5 | 821 | 92 | ||
| BC022948.1 | - | 2 - 3 | 317 | 92 | ||
| BC147484.1 | - | 4 - 5 | 667 | 9056231 | 92 | |
| BC147486.1 | - | 4 - 5 | 667 | 9056239 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnaja4
Genomic Map