Assay Details
| Gene Symbol: Fzd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021458.2 | NP_067433.1 | 3 - 4 | 844 | 106 | |
| GenBank mRNA | AK134261.1 | - | 3 - 4 | 780 | 106 | |
| AK160554.1 | - | 3 - 4 | 803 | 106 | ||
| BC050965.1 | - | 3 - 4 | 805 | 106 | ||
| U43205.1 | - | 2 - 3 | 693 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fzd3
Genomic Map