Assay Details
| Gene Symbol: Fzd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021458.2 | NP_067433.1 | 4 - 5 | 1860 | 63 | |
| GenBank mRNA | BC050965.1 | - | 4 - 5 | 1821 | 63 | |
| BC066186.1 | - | 4 - 5 | 1520 | 63 | ||
| BC103714.1 | - | 4 - 5 | 1520 | 63 | ||
| U43205.1 | - | 3 - 4 | 1709 | 63 | ||
More Information
Set Membership: |
|

Fzd3
Genomic Map