Assay Details
| Gene Symbol: Rps6kb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021485.2 | NP_067460.1 | 3 - 4 | 241 | 115 | |
| GenBank mRNA | AJ007938.1 | - | 3 - 4 | 241 | 115 | |
| AK014412.1 | - | 3 - 4 | 320 | 115 | ||
| AK090093.1 | - | 296 | 115 | |||
| AK156361.1 | - | 3 - 4 | 294 | 115 | ||
| BC051631.1 | - | 3 - 4 | 296 | 115 | ||
| BC066842.1 | - | 3 - 4 | 312 | 30468251 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rps6kb2
Genomic Map