| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500007606.mm.1 |
NM_001163487.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007606.mm.1 |
NM_001163487.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007606.mm.1 |
NM_001163488.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007606.mm.1 |
NM_001163488.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007606.mm.1 |
NM_021514.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007606.mm.1 |
NM_021514.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007606.mm.1 |
TC1500000969.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500007603.mm.1 |
NM_001163487.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007603.mm.1 |
NM_001163487.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007603.mm.1 |
TC1500000969.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500007604.mm.1 |
NM_021514.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007604.mm.1 |
NM_021514.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007604.mm.1 |
TC1500000969.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500007609.mm.1 |
NM_001163487.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007609.mm.1 |
NM_001163487.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007609.mm.1 |
NM_001163488.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007609.mm.1 |
NM_001163488.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007609.mm.1 |
NM_021514.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007609.mm.1 |
NM_021514.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007609.mm.1 |
TC1500000969.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500007605.mm.1 |
NM_021514.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007605.mm.1 |
NM_021514.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007605.mm.1 |
TC1500000969.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500007607.mm.1 |
NM_001163487.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007607.mm.1 |
NM_001163487.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007607.mm.1 |
NM_001163488.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007607.mm.1 |
NM_001163488.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007607.mm.1 |
NM_021514.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007607.mm.1 |
NM_021514.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007607.mm.1 |
TC1500000969.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500007608.mm.1 |
NM_001163487.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007608.mm.1 |
NM_001163487.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007608.mm.1 |
NM_001163488.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007608.mm.1 |
NM_001163488.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007608.mm.1 |
NM_021514.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007608.mm.1 |
NM_021514.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007608.mm.1 |
TC1500000969.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500007602.mm.1 |
NM_001163488.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007602.mm.1 |
NM_001163488.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500007602.mm.1 |
TC1500000969.mm.1 |
MTA TC |