Assay Details
| Gene Symbol: Stard3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021547.3 | NP_067522.1 | 1 - 2 | 142 | 80 | |
| GenBank mRNA | AK041538.1 | - | 1 - 2 | 98 | 80 | |
| AK150166.1 | - | 1 - 2 | 86 | 80 | ||
| AK150254.1 | - | 1 - 2 | 86 | 80 | ||
| AK155124.1 | - | 1 - 2 | 81 | 80 | ||
| AK160480.1 | - | 1 - 2 | 75 | 80 | ||
| BC003313.1 | - | 1 - 2 | 59 | 3155179 | 80 | |
| BC058727.1 | - | 1 - 2 | 79 | 80 | ||
| BY030024.1 | - | 1 - 2 | 143 | 80 | ||
| X82457.1 | - | 1 - 2 | 126 | 80 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK041538.1,AK155124.1,BC003313.1,NM_021547.3,X82457.1 transcripts.
Set Membership: |
|

Stard3
Genomic Map