Assay Details
| Gene Symbol: Jarid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205043.1 | NP_001191972.1 | 6 - 7 | 1507 | 67 | |
| NM_001205044.1 | NP_001191973.1 | 7 - 8 | 1180 | 67 | ||
| NM_021878.3 | NP_068678.1 | 7 - 8 | 1226 | 67 | ||
| GenBank mRNA | AK045214.1 | - | 6 - 7 | 1312 | 67 | |
| AK147226.1 | - | 6 - 7 | 1507 | 67 | ||
| AK164134.1 | - | 5 - 6 | 831 | 67 | ||
| AK216155.1 | - | 1 - 2 | 80 | 67 | ||
| BC026056.1 | - | 6 - 7 | 1414 | 67 | ||
| BC050129.1 | - | 6 - 7 | 1428 | 67 | ||
| BC051010.1 | - | 6 - 7 | 1410 | 67 | ||
| BC052444.1 | - | 7 - 8 | 1097 | 6406875 | 67 | |
| BC060695.1 | - | 7 - 8 | 1097 | 6406875 | 67 | |
| BC115758.1 | - | 1 - 2 | 111 | 67 | ||
| D31967.1 | - | 7 - 8 | 1116 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Jarid2
Genomic Map