Assay Details
| Gene Symbol: Tpm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253740.1 | NP_001240669.1 | 4 - 5 | 666 | 84 | |
| NM_001293748.1 | NP_001280677.2 | 5 - 6 | 649 | 84 | ||
| NM_001293749.1 | NP_001280678.1 | 4 - 5 | 666 | 84 | ||
| GenBank mRNA | AF317223.1 | - | 5 - 6 | 563 | 84 | |
| CX237327.1 | - | 4 - 5 | 488 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tpm3
Genomic Map