Assay Details
| Gene Symbol: Ddr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022563.2 | NP_072075.2 | 2 - 3 | 259 | 64 | |
| GenBank mRNA | BC138826.1 | - | 3 - 4 | 505 | 8861848 | 64 |
| BC138827.1 | - | 3 - 4 | 400 | 8861849 | 64 | |
| G49238.1 | - | 3 - 4 | 566 | 64 | ||
| X76505.1 | - | 3 - 4 | 566 | 64 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the X76505.1 transcripts.
Set Membership: |
|

Ddr2
Genomic Map