Assay Details
| Gene Symbol: Col4a3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164222.1 | NP_001157694.1 | 9 - 10 | 1445 | 73 | |
| NM_023420.2 | NP_075909.1 | 9 - 10 | 1445 | 73 | ||
| GenBank mRNA | AF232932.1 | - | 9 - 10 | 1459 | 73 | |
| AF232934.1 | - | 9 - 10 | 1459 | 73 | ||
| AK012989.1 | - | 9 - 10 | 1426 | 73 | ||
| AK030305.1 | - | 10 - 11 | 1353 | 73 | ||
| AK080958.1 | - | 9 - 10 | 1443 | 73 | ||
| BC016197.1 | - | 9 - 10 | 1408 | 4502118 | 73 | |
More Information
Set Membership: |
|

Col4a3bp
Genomic Map