Assay Details
| Gene Symbol: Col4a3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164222.1 | NP_001157694.1 | 15 - 16 | 2100 | 61 | |
| NM_023420.2 | NP_075909.1 | 16 - 17 | 2178 | 61 | ||
| GenBank mRNA | AF232932.1 | - | 16 - 17 | 2192 | 61 | |
| AF232934.1 | - | 15 - 16 | 2114 | 61 | ||
| AK012989.1 | - | 15 - 16 | 2081 | 61 | ||
| AK018103.1 | - | 4 - 5 | 469 | 61 | ||
| AK030305.1 | - | 16 - 17 | 2008 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Col4a3bp
Genomic Map