Assay Details
| Gene Symbol: Atp6v1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025494.3 | NP_079770.2 | 1 - 2 | 179 | 55 | |
| GenBank mRNA | AB088407.1 | - | 1 - 2 | 126 | 55 | |
| AK006323.1 | - | 1 - 2 | 130 | 55 | ||
| AK075779.1 | - | 1 - 2 | 151 | 55 | ||
| AK129007.1 | - | 1 - 2 | 139 | 55 | ||
| AK151237.1 | - | 1 - 2 | 140 | 55 | ||
| BB869068.1 | - | 1 - 2 | 180 | 55 | ||
| BC010217.1 | - | 1 - 2 | 127 | 3498388 | 55 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB088407.1,AK006323.1,AK075779.1,AK151237.1,BC010217.1,NM_025494.3 transcripts.
Set Membership: |
|

Atp6v1c1
Genomic Map