Assay Details
| Gene Symbol: Atp6v1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025494.3 | NP_079770.2 | 4 - 5 | 498 | 118 | |
| GenBank mRNA | AB088407.1 | - | 4 - 5 | 445 | 118 | |
| AK006323.1 | - | 4 - 5 | 449 | 118 | ||
| AK075779.1 | - | 4 - 5 | 470 | 118 | ||
| AK151237.1 | - | 4 - 5 | 459 | 118 | ||
| AK168914.1 | - | 4 - 5 | 449 | 118 | ||
| BC010217.1 | - | 4 - 5 | 446 | 3498388 | 118 | |
| U13839.1 | - | 4 - 5 | 441 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v1c1
Genomic Map