Assay Details
| Gene Symbol: Atp5e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025983.3 | NP_080259.1 | 1 - 2 | 140 | 80 | |
| GenBank mRNA | AK007809.1 | - | 1 - 2 | 129 | 80 | |
| AK010661.2 | - | 1 - 2 | 133 | 80 | ||
| AK012444.1 | - | 1 - 2 | 130 | 80 | ||
| AK131585.1 | - | 603 | 80 | |||
| AK160387.1 | - | 1 - 2 | 127 | 80 | ||
| AY341882.1 | - | 1 - 2 | 112 | 80 | ||
| BC024339.1 | - | 1 - 2 | 132 | 4981796 | 80 | |
| CJ322223.1 | - | 1 - 2 | 140 | 80 | ||
| W78532.1 | - | 1 - 2 | 115 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK007809.1, AK010661.2, AK012444.1, AK131585.1, AK160387.1, AY341882.1, BC024339.1, NM_025983.3 transcripts.
Set Membership: |
|

Atp5e
Genomic Map