Assay Details
| Gene Symbol: Abcg8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286005.1 | NP_001272934.1 | 10 - 11 | 1620 | 67 | |
| NM_026180.3 | NP_080456.1 | 11 - 12 | 1872 | 67 | ||
| NR_104382.1 | 11 - 12 | 1869 | 67 | |||
| GenBank mRNA | AF324495.1 | - | 11 - 12 | 1862 | 67 | |
| AK004871.1 | - | 11 - 12 | 1829 | 67 | ||
| AK008188.1 | - | 1 - 2 | 45 | 67 | ||
| AK050938.1 | - | 10 - 11 | 1622 | 67 | ||
| AY196216.1 | - | 11 - 12 | 1859 | 67 | ||
| BC138484.1 | - | 11 - 12 | 1836 | 8861504 | 67 | |
| BC145293.1 | - | 11 - 12 | 1836 | 9053847 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abcg8
Genomic Map