Assay Details
| Gene Symbol: Snx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026386.1 | NP_080662.1 | 1 - 2 | 123 | 69 | |
| GenBank mRNA | AK002692.1 | - | 1 - 2 | 123 | 69 | |
| AK005470.1 | - | 1 - 2 | 135 | 69 | ||
| AK010572.1 | - | 1 - 2 | 120 | 69 | ||
| AK075929.1 | - | 1 - 2 | 120 | 69 | ||
| AK145840.1 | - | 1 - 2 | 174 | 69 | ||
| AK153525.1 | - | 1 - 2 | 120 | 69 | ||
| BC006960.1 | - | 1 - 2 | 123 | 2812557 | 69 | |
| CT010229.1 | - | 1 - 2 | 108 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Snx2
Genomic Map