Assay Details
| Gene Symbol: Sfxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027324.5 | NP_081600.1 | 1 - 2 | 91 | 69 | |
| GenBank mRNA | AF325260.1 | - | 1 - 2 | 69 | 69 | |
| AK012650.1 | - | 1 - 2 | 90 | 69 | ||
| BC005743.1 | - | 1 - 2 | 48 | 3599636 | 69 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF325260.1,BC005743.1,NM_027324.5 transcripts.
Set Membership: |
|

Sfxn1
Genomic Map