Assay Details
| Gene Symbol: Coasy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF521095.1 | - | 5 - 6 | 1271 | 111 | |
| AK004878.1 | - | 5 - 6 | 1331 | 111 | ||
| AK159405.1 | - | 5 - 6 | 1320 | 111 | ||
| AK159912.1 | - | 4 - 5 | 1620 | 111 | ||
| AK166894.1 | - | 4 - 5 | 1614 | 111 | ||
| AK170330.1 | - | 4 - 5 | 1629 | 111 | ||
| BC020046.1 | - | 4 - 5 | 741 | 111 | ||
| BC064753.1 | - | 4 - 5 | 1259 | 111 | ||
More Information
Set Membership: |
|

Coasy
Genomic Map