Assay Details
| Gene Symbol: Coasy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF521095.1 | - | 8 - 9 | 1522 | 80 | |
| AK004878.1 | - | 8 - 9 | 1582 | 80 | ||
| AK159405.1 | - | 8 - 9 | 1571 | 80 | ||
| AK159912.1 | - | 7 - 8 | 1871 | 80 | ||
| AK166894.1 | - | 7 - 8 | 1865 | 80 | ||
| AK170330.1 | - | 7 - 8 | 1880 | 80 | ||
| BC020046.1 | - | 7 - 8 | 992 | 80 | ||
| BC064753.1 | - | 7 - 8 | 1510 | 80 | ||
More Information
Set Membership: |
|

Coasy
Genomic Map