Assay Details
| Gene Symbol: Abcg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030239.2 | NP_084515.2 | 4 - 5 | 571 | 88 | |
| GenBank mRNA | AF324242.1 | - | 4 - 5 | 523 | 88 | |
| AK030831.1 | - | 4 - 5 | 573 | 88 | ||
| AK037614.1 | - | 4 - 5 | 487 | 88 | ||
| AK051880.1 | - | 4 - 5 | 537 | 88 | ||
| AK156626.1 | - | 4 - 5 | 539 | 88 | ||
| BC138772.1 | - | 4 - 5 | 552 | 8861794 | 88 | |
| BC138773.1 | - | 4 - 5 | 551 | 8861795 | 88 | |
More Information
Set Membership: |
|

Abcg3
Genomic Map