Assay Details
| Gene Symbol: Acox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030721.2 | NP_109646.2 | 9 - 10 | 1061 | 63 | |
| GenBank mRNA | AJ278430.1 | - | 9 - 10 | 1094 | 63 | |
| AK028818.1 | - | 9 - 10 | 1482 | 63 | ||
| AK053960.1 | - | 9 - 10 | 1058 | 63 | ||
| AK128922.1 | - | 9 - 10 | 1021 | 63 | ||
| AK154112.1 | - | 8 - 9 | 1152 | 63 | ||
| AK171598.1 | - | 1044 | 63 | |||
| BC044725.2 | - | 9 - 10 | 1081 | 5374281 | 63 | |
| BC055019.1 | - | 9 - 10 | 1061 | 4162223 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acox3
Genomic Map