Assay Details
| Gene Symbol: Cd247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113391.2 | NP_001106862.1 | 2 - 3 | 312 | 59 | |
| NM_001113392.2 | NP_001106863.1 | 2 - 3 | 312 | 59 | ||
| NM_001113393.2 | NP_001106864.2 | 2 - 3 | 312 | 59 | ||
| NM_031162.4 | NP_112439.1 | 2 - 3 | 312 | 59 | ||
| NR_103716.1 | 2 - 3 | 312 | 59 | |||
| GenBank mRNA | AK017904.1 | - | 2 - 3 | 275 | 59 | |
| AK088087.1 | - | 2 - 3 | 240 | 59 | ||
| AK138774.1 | - | 2 - 3 | 313 | 59 | ||
| BC052824.1 | - | 2 - 3 | 233 | 30074812 | 59 | |
| J04967.1 | - | 2 - 3 | 304 | 59 | ||
| L03353.1 | - | 2 - 3 | 228 | 59 | ||
| M33158.1 | - | 2 - 3 | 302 | 59 | ||
| S51932.1 | - | 2 - 3 | 228 | 59 | ||
| U56258.1 | - | 2 - 3 | 228 | 59 | ||
| X84237.1 | - | 2 - 3 | 228 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd247
Genomic Map