Assay Details
| Gene Symbol: Kcnn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032397.2 | NP_115773.2 | 5 - 6 | 1210 | 102 | |
| GenBank mRNA | AF116525.1 | - | 4 - 5 | 1238 | 102 | |
| AF357239.1 | - | 5 - 6 | 1166 | 102 | ||
| AK041033.1 | - | 5 - 6 | 1211 | 102 | ||
| AK043611.1 | - | 5 - 6 | 1180 | 102 | ||
| AY258143.1 | - | 4 - 5 | 1049 | 102 | ||
| BC090622.1 | - | 6 - 7 | 1553 | 6832359 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnn1
Genomic Map